Learn More
Bellco Glass, Inc. The Dounce tissue homogenizer was designed for disruption of whole cells leaving cell nuclei undamaged. Two pestles are supplied with each complete unit. Large pestle is used for the initial sample reduction. The small pestle is used to form the final hom
SDP
Supplier:  Bellco Glass, Inc. 198410100
Encompass_Preferred
The Dounce tissue homogenizer was designed for disruption of whole cells leaving cell nuclei undamaged. Two pestles are supplied with each complete unit. Large pestle is used for the initial sample reduction. The small pestle is used to form the final hom
Catalog No. 50-194-5208