Learn More
Bellco Glass, Inc. The Dounce tissue homogenizer was designed for disruption of whole cells leaving cell nuclei undamaged. Two pestles are supplied with each complete unit. Large pestle is used for the initial sample reduction. The small pestle is used to form the final hom
SDP
Catalog No. 501945207
Encompass_Preferred
The Dounce tissue homogenizer was designed for disruption of whole cells leaving cell nuclei undamaged. Two pestles are supplied with each complete unit. Large pestle is used for the initial sample reduction. The small pestle is used to form the final hom
Catalog No. 50-194-5207 Supplier Bellco Glass, Inc. Supplier No. 198410040
The Dounce tissue homogenizer was designed for disruption of whole cells leaving cell nuclei undamaged. Two pestles are supplied with each complete unit. Large pestle is used for the initial sample reduction. The small pestle is used to form the final hom