Learn More
Bellco Glass, Inc. Dounce Homogenizer,7mL
SDP
Catalog No. 501945205
Encompass_Preferred
Dounce Homogenizer Working Capacity: 7mL All glass construction Two pestles are supplied with ea unit *large pestle is used for the initial sample reduction *small pestle is used to form the final homogenate Large pestle clearance: 0028-0047" Small pestle clearance: .0008-0022" Tube Overall: 125mm Desiged primarly for cellular work where the nuclues remains intact after homogenization Qty: 1 ea. Manufactured from borosilicate glass 3.3Designed primarily for cellular work where the nucleus remains intact after homogenization.All-glass constructionTwo pestles are supplied with each complete unitLarge clearance pestle is used for the initial sample reductionSmall clearance pestle is used to form the final homogenateReplacement components are available and completely interchangeable
Catalog No. 50-194-5205 Supplier Bellco Glass, Inc. Supplier No. 198410007
Dounce Homogenizer Working Capacity: 7mL All glass construction Two pestles are supplied with ea unit *large pestle is used for the initial sample reduction *small pestle is used to form the final homogenate Large pestle clearance: 0028-0047" Small pestle clearance: .0008-0022" Tube Overall: 125mm Desiged primarly for cellular work where the nuclues remains intact after homogenization Qty: 1 ea. Manufactured from borosilicate glass 3.3Designed primarily for cellular work where the nucleus remains intact after homogenization.All-glass constructionTwo pestles are supplied with each complete unitLarge clearance pestle is used for the initial sample reductionSmall clearance pestle is used to form the final homogenateReplacement components are available and completely interchangeable