Learn More
Andwin Scientific DOUNCE HOMOGENIZER Capacity: 2 mL; Tube Length:60 mm, Large Pestle Clearance:0.8890.127mm
SDP
Catalog No. NC9357430
Encompass_Preferred
Capacity: 2 mL; Tube Length:60 mm, Large Pestle Clearance:0.8890.127mm; Small Pestle Clearance:0.0130.064 mm; For cellular work where the nucleus remains intact after homogenization. Borosilicate glass construction. One tube and two pestles are supplied with each tissue grinder. The large clearance pestle is used for initial sample reduction. The small clearance pestle forms the final homogenate.
Catalog No. NC9357430 Supplier Andwin Scientific Supplier No. KT8853000002
Capacity: 2 mL; Tube Length:60 mm, Large Pestle Clearance:0.8890.127mm; Small Pestle Clearance:0.0130.064 mm; For cellular work where the nucleus remains intact after homogenization. Borosilicate glass construction. One tube and two pestles are supplied with each tissue grinder. The large clearance pestle is used for initial sample reduction. The small clearance pestle forms the final homogenate.