Learn More
Electron Microscopy Sciences Dounce Tissue Grinder, 15 ml, 22 mmx 94 mm Mortor, 210 mm OAL
SDP
Catalog No. 50365335
Encompass_Preferred
This grinderis ideal for soft tissue and will retain a high percentage of cell nuclei and mitochondria in soft tissues from cell cultures. When in use, the pestle ball is encircled in liquid, which avoids heat buildup by reducing friction. The initial grinding is performed using the "loose" pestle. The grinding process is completed using the "tight" pestle. This grinder is used by moving the pestle up and down, and work best with cell suspensions or very soft tissues. The mortar has a large reservoir and pouring lip and is supplied with "loose” and "tight” pestles. Made of Wheaton 33 low extractable borosilicate glass. Autoclavable.
Size: 15 ml
Mortor: 22 mmx 94 mm
OAL: 210 mm
Catalog No. 50-365-335 Supplier Electron Microscopy Sciences Supplier No. 6479015
This grinderis ideal for soft tissue and will retain a high percentage of cell nuclei and mitochondria in soft tissues from cell cultures. When in use, the pestle ball is encircled in liquid, which avoids heat buildup by reducing friction. The initial grinding is performed using the "loose" pestle. The grinding process is completed using the "tight" pestle. This grinder is used by moving the pestle up and down, and work best with cell suspensions or very soft tissues. The mortar has a large reservoir and pouring lip and is supplied with "loose” and "tight” pestles. Made of Wheaton 33 low extractable borosilicate glass. Autoclavable.
Size: 15 ml
Mortor: 22 mmx 94 mm
OAL: 210 mm