Learn More
Wilmad Labglass Dounce Tissue Grinders, 2 mL
Catalog No. NC1880753
Encompass_Preferred
Dounce tissue grinders are made from smooth surfaced borosilicate glass to eliminate glass particles in the reduced sample. They are useful in enzyme studies and recommended where heat build-up should be avoided.
Catalog No. NC1880753 Supplier Wilmad Labglass Supplier No. LG10645090
Dounce tissue grinders are made from smooth surfaced borosilicate glass to eliminate glass particles in the reduced sample. They are useful in enzyme studies and recommended where heat build-up should be avoided.