Learn More
Medchemexpress LLC HY-18332C 5mg Medchemexpress, DOV-216,303 (Free Base) CAS:66504-40-3 Purity:>98%
SDP
Catalog No. 501932812
Encompass_Preferred
Medchemexpress, HY-18332C 5mg DOV-216,303 (Free Base) CAS:66504-40-3 DOV-216,303 (Free Base) is a potent triple serotonin , norepinephrine , and dopamine reuptake inhibitor, with IC 50 values of 14 nM, 20 nM and 78 nM for hSERT, hNET and hDAT, respectively [1] . Has antidepressant-like effects and increases monoamine release in the prefrontal cortex of olfactory bulbectomized (OBX) rats [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2812 Supplier Medchemexpress LLC Supplier No. HY18332C5MG
Medchemexpress, HY-18332C 5mg DOV-216,303 (Free Base) CAS:66504-40-3 DOV-216,303 (Free Base) is a potent triple serotonin , norepinephrine , and dopamine reuptake inhibitor, with IC 50 values of 14 nM, 20 nM and 78 nM for hSERT, hNET and hDAT, respectively [1] . Has antidepressant-like effects and increases monoamine release in the prefrontal cortex of olfactory bulbectomized (OBX) rats [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.