Learn More
Description
- For metastatic ovarian cancers
- Used in studies to focus on cellular migration and invasion
- Responsive to vascular endothelial growth factor (VEGF), express estrogen receptor and matrilysin (MMP-7)
- For my metastatic ovarian cancer studies
Specifications
Specifications
| Content And Storage | Store in liquid nitrogen, Can be cultured at least 10 passages after initial thawing without significantly affecting cell marker expression and functionality |
| Quantity | 1 x 10&sup6; Cells/Vial |
| For Use With (Application) | Cell Culture |
| Sample Type | Cancer cells |
| Form | Liquid |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.