Learn More
Selleck Chemical LLC Dovitinib (TKI-258) 10mM/1mL 405169-16-6 CHIR-258
SDP
Catalog No. 501366111
Encompass_Preferred
Dovitinib (TKI258, CHIR258) is a multitargeted RTK inhibitor, mostly for class III (FLT3/c-Kit) with IC50 of 1 nM/2 nM, also potent to class IV (FGFR1/3) and class V (VEGFR1-4) RTKs with IC50 of 8-13 nM, less potent to InsR, EGFR, c-Met, EphA2, Tie2, IGF-1R and HER2 in cell-free assays. Phase 4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6111 Supplier Selleck Chemical LLC Supplier No. S101810MM/1ML
Dovitinib (TKI258, CHIR258) is a multitargeted RTK inhibitor, mostly for class III (FLT3/c-Kit) with IC50 of 1 nM/2 nM, also potent to class IV (FGFR1/3) and class V (VEGFR1-4) RTKs with IC50 of 8-13 nM, less potent to InsR, EGFR, c-Met, EphA2, Tie2, IGF-1R and HER2 in cell-free assays. Phase 4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.