Learn More
Selleck Chemical LLC Dovitinib
SDP
Catalog No. 5000003887
Encompass_Preferred
Dovitinib (TKI258) Lactate monohydrate is the Lactate of Dovitinib which is a multitargeted RTK inhibitor mostly for class III (FLT3/c-Kit) with IC50 of 1 nM/2 nM also potent to class IV (FGFR1/3) and class V (VEGFR1-4) RTKs with IC50 of 8-13 nM less potent to InsR EGFR c-Met EphA2 Tie2 IGFR1 and HER2 Phase 4
Catalog No. 50-000-03887 Supplier Selleck Chemical LLC Supplier No. S776510MG
Dovitinib (TKI258) Lactate monohydrate is the Lactate of Dovitinib which is a multitargeted RTK inhibitor mostly for class III (FLT3/c-Kit) with IC50 of 1 nM/2 nM also potent to class IV (FGFR1/3) and class V (VEGFR1-4) RTKs with IC50 of 8-13 nM less potent to InsR EGFR c-Met EphA2 Tie2 IGFR1 and HER2 Phase 4