Learn More
TARGETMOL CHEMICALS INC DOVITINIB LACTATE HYDRAT 25MG
SDP
Catalog No. 502579354
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dovitinib lactate hydrate (TKI258) is the Lactate of Dovitinib which is a multitargeted RTK inhibitor mostly for class III (FLT3/c-Kit) with IC50 of 1 nM/2 nM also potent to class IV (FGFR1/3) and class V (VEGFR1-4) RTKs with IC50 of 8-13 nM less potent to InsR EGFR c-Met EphA2 Tie2 IGFR1 and HER2. Phase 4. purity: 99%
Catalog No. 50-257-9354 Supplier TARGETMOL CHEMICALS INC Supplier No. T647925MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dovitinib lactate hydrate (TKI258) is the Lactate of Dovitinib which is a multitargeted RTK inhibitor mostly for class III (FLT3/c-Kit) with IC50 of 1 nM/2 nM also potent to class IV (FGFR1/3) and class V (VEGFR1-4) RTKs with IC50 of 8-13 nM less potent to InsR EGFR c-Met EphA2 Tie2 IGFR1 and HER2. Phase 4. purity: 99%