Learn More
Apexbio Technology LLC Dovitinib (TKI-258, CHIR-258), 5mg. Cas: 405169-16-6 MFCD: MFCD10565680. Multitargeted RTK inhibitor
SDP
Supplier:  Apexbio Technology LLC A21685
Encompass_Preferred
Dovitinib (TKI258, CHIR-258) is a multitargeted receptor tyrosine kinase inhibitor against FLT3, KIT, FGFR, VEGFR, PDGFR and PDGFR with IC50 of 1 nM, 2 nM, 8-9 nM, 8-13 nM , 210 nM and 27nM, respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4734