Learn More
Krayden DOW CORNING 3-4680 SIL GEL KIT
Catalog No. NC0676382
Encompass_Preferred
Dow Corning 3-4680 Silicone Gel is two-part, fast RT cure gel with low viscosity which allows good flow under components. 2-part, transparent blue, 1:1 mix ratio, fast room-temperature-cure gel.
Catalog No. NC0676382 Supplier Krayden Supplier No. DC4027868
Dow Corning 3-4680 Silicone Gel is two-part, fast RT cure gel with low viscosity which allows good flow under components. 2-part, transparent blue, 1:1 mix ratio, fast room-temperature-cure gel.