Learn More
Andwin Scientific DOW CORNING VAC GREASE 150G EA
SDP
Catalog No. 50727537
Encompass
The Dow Corning Vacuum Grease is a silicone lubricant for glass-to-glass joints and glass-to-rubber connections. This product features resistance against chemicals and stability in the heat. It is packaged in a tube and is sold in a quantity of 150 grams.
Catalog No. 50-727-537 Supplier Andwin Scientific Supplier No. 60705
The Dow Corning Vacuum Grease is a silicone lubricant for glass-to-glass joints and glass-to-rubber connections. This product features resistance against chemicals and stability in the heat. It is packaged in a tube and is sold in a quantity of 150 grams.