Learn More
Fine Science Tools Dowell Scissors, Angled Up, 0.1 mm Tip Dia, 11.5 cm
SDP
Catalog No. NC1706221
Encompass
Tip Shape: Angled Up/ Tips: Sharp/ Cutting Edge: 10mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No
Catalog No. NC1706221 Supplier Fine Science Tools Supplier No. 1504211
Tip Shape: Angled Up/ Tips: Sharp/ Cutting Edge: 10mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No