Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 1X2 100-200 (Cl)

Catalog No. p-7046415
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1425418 50 g
AAL1425430 250 g
AAL142540B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1425418 Supplier Thermo Scientific Chemicals Supplier No. L1425418
Only null left
Add to Cart

CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9085-42-1
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132715
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
IUPAC Name 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
SMILES [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1

Specifications

Odor Amine-like
Quantity 50 g
Chemical Name or Material Dowex™ 1X2 100-200 (Cl)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.