Learn More
MilliporeSigma™ Supelco™ Dowex™ 1X4 Chloride Form, 200-400 mesh
Shop All MilliporeSigma Supelco ProductsDescription
- AmberChrom™ 1X4 is a strongly basic, type I, anion exchange resin featuring a styrene-divinylbenzene gel matrix.
- It is suitable for LPLC and meets the requirements of FDA Food Additive Regulation 21 CFR 173.25.
- Matrix: Styrene-divinylbenzene (gel)
- Active Group: Quaternary ammonium
- Cross-linkage: 4%
- Particle Size: 200-400 mesh
- Capacity: 1.0 meq/mL
- Operating pH: 0 to 14
- Maximum Temperature: 99°C (Cl- form), 66°C (OH- form)
- Moisture: 55% to 63%
- Form: Beads
Specifications
Specifications
| Particle Size | 200/400 Mesh |
| Technique | Chromatography |
| Packaging | Poly Bottle |
| pH | 0 to 14 |
| Quantity | 500 g |
| Product Line | DOWEX |
| Material | Styrene-divinylbenzene |
| Capacity (Metric) | 1.0 meq/mL capacity |
| MDL Number | MFCD00146764 |
| CAS | 69011-19-4 |
DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.