Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Dowex™ 1X8 Chloride Form, 20-50 mesh

Catalog No. 111015194 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5194 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5194 Supplier MilliporeSigma™ Supelco™ Supplier No. 44324100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strongly basic

Dowex™ 1X8 is a strongly basic anion exchange resin which is used as an absorbent in solid phase extraction.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 20/50 Mesh
pH 0 to 14
Quantity 100 g
Product Line DOWEX
Capacity (Metric) 1.2 meq/mL by wetted bed volume
MDL Number MFCD00132718
CAS 69011-19-4

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.