Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Dowex™ 1X8 Chloride Form, 50-100 mesh

Catalog No. 111015200 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5200 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5200 Supplier MilliporeSigma™ Supelco™ Supplier No. 217417500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This ion-exchange resin has not been specially processed nor cleaned. It is suggested that it be treated to a suitable preliminary elution and wash.

Dowex™ 1X8 is a strongly basic anion exchange resin which is used as an absorbent in solid phase extraction.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 50/100 Mesh
Packaging Poly Bottle
pH 0 to 14
Quantity 500 g
Product Line DOWEX
Capacity (Metric) 1.2 meq/mL by wetted bed volume
MDL Number MFCD00132718
CAS 69011-19-4

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.