Learn More
MilliporeSigma™ Supelco™ Dowex™ 1X8 Formate Form, 200-400 mesh
Catalog No. 111006049 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2.5 kg
Catalog No. Quantity
11-100-6049 2.5 kg
1 options
Catalog No. 11-100-6049 Supplier MilliporeSigma™ Supelco™ Supplier No. 13858U
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Dowex™ 1X8, a strongly basic anion exchange resin, has been used as an absorbent in solid phase extraction.
Dowex™ 1X8 formate form was used to remove neutral monosaccharides and hexosamines from hydrolysates using ion-exchange chromatography. Dowex™ 1X8 formate form was used to purify organic acids from lyophilized fractions.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 200/400 Mesh
Quantity 2.5 kg
Product Line DOWEX
Capacity (Metric) 1.0 meq/mL
DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow