Learn More
Dowex™ 50WX4 200-400 (H)
Description
Dowex™ 50WX4 200-400 (H) is used to investigate the effect of particle size on drug loading and release profiles. Dowex™ 50WX4 hydrogen form when treated in boiling water converted D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) to free aldehyde D,L-glyceraldehyde 3-phosphate. It is a useful column packing material, used in anion exchange chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dowex™ 50WX4 200-400 (H) is used to investigate the effect of particle size on drug loading and release profiles. Dowex™ 50WX4 hydrogen form when treated in boiling water converted D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) to free aldehyde D,L-glyceraldehyde 3-phosphate. It is a useful column packing material, used in anion exchange chromatography.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 50 g |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Dowex™ 50WX4 200-400 (H) |
Safety and Handling
| Hazard Category | H318 |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.