Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 50X4-400, ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
11113-61-4
MFCD00132725
11113-61-4
MFCD00132725
Particle Size Particle size analysis: on 100 mesh (cumulative): <15%, through 325 mesh: <15%
Packaging Plastic bottle
Water Volume Capacity wet volume capacity, H form (meq/mL):> 1.1
Water Retention Capacity H form: 64 to 72%
Quantity 2.5kg
Physical Form Beads
Chemical Name or Material Dowex™ 50WX4