Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 50X8-200, ion-exchange resin, Thermo Scientific™

Catalog No. AC203071000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC203071000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC203071000 Supplier Thermo Scientific Chemicals Supplier No. 203071000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin

Specifications

Physical Form Beads
Particle Size Particle size analysis: on 60 mesh (cumulative): <15%, through 170 mesh: <15%
Volume Total Capacity wet volume capacity, H form (meq/mL):> 1.7
Water Retention Capacity H form: 50 to 58%
Quantity 100 g
Chemical Name or Material Dowex(r) 50X8-200
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.