Learn More
Dowex™ G26 hydrogen form hydrogen form, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 210235-42-0 |
| Product Line | DOWEX |
| Density | 1.22 g/mL |
| Type | Organic Standard |
| Quantity | 5 kg |
DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.