Learn More
MilliporeSigma™ Supelco™ Dowex Marathon C, Hydrogen Form, 23-27 mesh
Catalog No. 111002462 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 g
Catalog No. Quantity
11-100-2462 250 g
1 options
Catalog No. 11-100-2462 Supplier MilliporeSigma™ Supelco™ Supplier No. 433950250G
This resin has not been specially processed nor cleaned. It is suggested that it be subjected to a suitable preliminary elution and wash.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 23/27 Mesh
Packaging Fiber Drum
Quantity 250 g
Product Line Marathon
Capacity (Metric) 1.8 meq/mL by wetted bed volume
MDL Number MFCD00212649
Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow