Learn More
Crescent Chemical Co Inc DOWEX 1X2 (200-400 MESH)
SDP
Catalog No. 50248508
Encompass_Preferred
DOWEX 1X2 (200-400 mesh) analytical grade Anion exchanger of type I, strongly basic. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 41031.02
Catalog No. 50-248-508 Supplier Crescent Chemical Co Inc Supplier No. SE41031.02
DOWEX 1X2 (200-400 mesh) analytical grade Anion exchanger of type I, strongly basic. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 41031.02