Learn More
Crescent Chemical Co Inc DOWEX 1X8 (100-200 MESH)
SDP
Catalog No. 50248520
Encompass_Preferred
DOWEX 1X8 (100-200 mesh) analytical grade Anion exchanger of type I, strongly basic. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 41101.02
Catalog No. 50-248-520 Supplier Crescent Chemical Co Inc Supplier No. SE41101.02
DOWEX 1X8 (100-200 mesh) analytical grade Anion exchanger of type I, strongly basic. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 41101.02