Learn More
Crescent Chemical Co Inc DOWEX 1X8 (100-200 MESH)
Supplier: Crescent Chemical Co Inc SE41101.01
DOWEX 1X8 (100-200 mesh) analytical grade Anion exchanger of type I, strongly basic. Size - 100G Storage Conditions - +15 C TO +30 C Catalog Number - 41101.01