Learn More
Crescent Chemical Co Inc DOWEX 1X8 (20-50 MESH)
SDP
Catalog No. 50248512
Encompass_Preferred
DOWEX 1X8 (20-50 mesh) analytical grade Anion exchanger of type I, strongly basic. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 41081.04
Catalog No. 50-248-512 Supplier Crescent Chemical Co Inc Supplier No. SE41081.04
DOWEX 1X8 (20-50 mesh) analytical grade Anion exchanger of type I, strongly basic. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 41081.04