Learn More
Crescent Chemical Co Inc DOWEX 1X8 (20-50 MESH)
SDP
Supplier:  Crescent Chemical Co Inc SE41080.04
Encompass_Preferred
DOWEX 1X8 (20-50 mesh) pract. Anion exchanger of type I, strongly basic. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 41080.04
Catalog No. 50-248-510