Learn More
Crescent Chemical Co Inc DOWEX 50 WX8 (100-200 MESH)
SDP
Supplier:  Crescent Chemical Co Inc SE41621.02
Encompass_Preferred
DOWEX 50 WX8 (100-200 mesh) analytical grade Cation exchanger, strongly acidic. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 41621.02
Catalog No. 50-248-548