Learn More
Crescent Chemical Co Inc DOWEX 50 WX8 (100-200 MESH)
SDP
Catalog No. 50248547
Encompass_Preferred
DOWEX 50 WX8 (100-200 mesh) analytical grade Cation exchanger, strongly acidic. Size - 100G Storage Conditions - +15 C TO +30 C Catalog Number - 41621.01
Catalog No. 50-248-547 Supplier Crescent Chemical Co Inc Supplier No. SE41621.01
DOWEX 50 WX8 (100-200 mesh) analytical grade Cation exchanger, strongly acidic. Size - 100G Storage Conditions - +15 C TO +30 C Catalog Number - 41621.01