Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex(R), 50WX8-200, Ion Exchange Resin, Dowex-50W-hydrogen, Spectrum™ Chemical
SDP

Catalog No. 18602322 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-322 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-602-322 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DWX65100GM

missing translation for 'validMainframeMsg'

DWX65, 11119-67-8

Spectrum™ Chemical Dowex, 50WX8-200, Ion Exchange Resin, Dowex-50W-hydrogen is a polymer containing sulfonate functional groups or containing sulfonic acid that comes from polystyrene. Ion exchange resins are used in different separation, purification, and decontamination processes. The most common examples are water softening and water purification. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChromTM 50WX8 Ion Exchange Resin

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.