Learn More
EZKEM Dowfax 2A1 1L Cubitainer
SDP
Catalog No. NC1451510
Encompass_Preferred
EZkem’s 1L Dowfax 2A1 surfactant solution is used to help liquids flow through tubing easier and to help facilitate mixing. Used on Alpkem and OI Analytical FSIV, FSIII and FS3000 autoanalyzers, flow injection analyzers and segmented flow analyzers.
Catalog No. NC1451510 Supplier EZKEM Supplier No. E060064
EZkem’s 1L Dowfax 2A1 surfactant solution is used to help liquids flow through tubing easier and to help facilitate mixing. Used on Alpkem and OI Analytical FSIV, FSIII and FS3000 autoanalyzers, flow injection analyzers and segmented flow analyzers.