Learn More
EZKEM Dowfax 2A1 125 ml
SDP
Catalog No. NC1122586
Encompass_Preferred
EZkem’s 125 mL Dowfax 2A1 surfactant solution is used to help liquids flow through tubing easier and to help facilitate mixing. Used on Alpkem and OI Analytical FSIV, FSIII and FS3000 autoanalyzers, flow injection analyzers and segmented flow analyzers.
Catalog No. NC1122586 Supplier EZKEM Supplier No. A000080
EZkem’s 125 mL Dowfax 2A1 surfactant solution is used to help liquids flow through tubing easier and to help facilitate mixing. Used on Alpkem and OI Analytical FSIV, FSIII and FS3000 autoanalyzers, flow injection analyzers and segmented flow analyzers.