Learn More
Astoria Pacific International DOWFAZ 2A1 32 OZ
SDP
Catalog No. NC0955177
Encompass_Preferred
This item has a minimum qty of 4 per supplier requirements.
Color Amber; Container Type Bottle; For Use With (Application) Provides excellent solubility and stability in Acidic, Alkaline, bleach and other oxiding systems and flow analyzers; Format Liquid; pH Range 8 to 10.5.
This item is not returnable. View return policy
Catalog No. NC0955177 Supplier Astoria Pacific International Supplier No. 90072032
This item has a minimum qty of 4 per supplier requirements.
Color Amber; Container Type Bottle; For Use With (Application) Provides excellent solubility and stability in Acidic, Alkaline, bleach and other oxiding systems and flow analyzers; Format Liquid; pH Range 8 to 10.5.