Learn More
Astoria Pacific International DOWFAZ 2A1 4 OZ
SDP
Catalog No. NC0955176
Encompass_Preferred
This item has a minimum qty of 12 per supplier requirements.
Color Amber; Container Type Bottle; For Use With (Application) Provides excellent solubility and stability in Acidic, Alkaline, bleach and other oxiding systems and flow analyzers; Format Liquid; pH Range 8 to 10.5.
This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. NC0955176 Supplier Astoria Pacific International Supplier No. 90072004
This item has a minimum qty of 12 per supplier requirements.
Color Amber; Container Type Bottle; For Use With (Application) Provides excellent solubility and stability in Acidic, Alkaline, bleach and other oxiding systems and flow analyzers; Format Liquid; pH Range 8 to 10.5.
This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.