Learn More
Hubbard Hall Dowfrost Concentrate, 5 gal
SDP
Catalog No. NC9258176
Encompass
Dowfrost Concentrate is widely used for secondary heating and cooling applications. It is useful in applications where toxicity is a concern because of its low acute oral toxicity. It is used where contact with drinking water is possible and in food processing applications where accidental contact with food or beverage products could occur.
Catalog No. NC9258176 Supplier Hubbard Hall Supplier No. 46110035 GAL
Dowfrost Concentrate is widely used for secondary heating and cooling applications. It is useful in applications where toxicity is a concern because of its low acute oral toxicity. It is used where contact with drinking water is possible and in food processing applications where accidental contact with food or beverage products could occur.