Learn More
Chempoint Com DOWTHERM™ SR-1 Heat Transfer Fluid, Dyed, 5G pail
Catalog No. NC0239140
Encompass
A formulation of ethylene glycol and a specially designed package of industrial corrosion inhibitors with operating temperature ranges of -60°F to 250°F. The fluid is dyed fluorescent pink for leak detection purposes. Heat transfer fluid. For non-evaporative closed loop systems.
Catalog No. NC0239140 Supplier Chempoint Com Supplier No. 0124558
A formulation of ethylene glycol and a specially designed package of industrial corrosion inhibitors with operating temperature ranges of -60°F to 250°F. The fluid is dyed fluorescent pink for leak detection purposes. Heat transfer fluid. For non-evaporative closed loop systems.