Learn More
Medchemexpress LLC Doxapram-d5 hydrochloride | 00-00-0 | 99.0% | 420.00 | C24H26D5ClN2O2 | 5 MG
SDP
Catalog No. 5000262903
Encompass_Preferred
Doxapram-d5 hydrochloride is the deuterium-labeled isotopologue of doxapram hydrochloride supplied as a solid 5 mg vial. It is provided as a high-purity analytical reference for use as an internal standard and a pharmacological research reagent in studies of respiratory stimulation and ion channel modulation. The material is intended for laboratory research and analytical quantification.
Catalog No. 50-002-62903 Supplier Medchemexpress LLC Supplier No. HY129146S5MG
Doxapram-d5 hydrochloride is the deuterium-labeled isotopologue of doxapram hydrochloride supplied as a solid 5 mg vial. It is provided as a high-purity analytical reference for use as an internal standard and a pharmacological research reagent in studies of respiratory stimulation and ion channel modulation. The material is intended for laboratory research and analytical quantification.