Learn More
Medchemexpress LLC Doxapram-d8 | 309-29-5 | 99.5% | 1 MG
SDP
Catalog No. 5000407843
Encompass_Preferred
Doxapram-d8 is deuterated labeled Doxapram, which is a respiratory stimulant. It increases breathing rate and depth by acting on the brain's respiratory centers and peripheral chemoreceptors. Doxapram inhibits TASK-1, TASK-3, TASK-1/TASK-3 heterodimeric channel function with EC50s of 410 nM, 37 μM, 9 μM, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07843 Supplier Medchemexpress LLC Supplier No. HYB0551S1MG
Doxapram-d8 is deuterated labeled Doxapram, which is a respiratory stimulant. It increases breathing rate and depth by acting on the brain's respiratory centers and peripheral chemoreceptors. Doxapram inhibits TASK-1, TASK-3, TASK-1/TASK-3 heterodimeric channel function with EC50s of 410 nM, 37 μM, 9 μM, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.