Learn More
Medchemexpress LLC Doxapram-d8 | 309-29-5 | 99.5% | 386.56 g/mol | C24H22D8N2O2 | 10 MG
SDP
Catalog No. 5000298918
Encompass_Preferred
Doxapram-d8 is a deuterium-labeled form of doxapram provided as a research standard for analytical and pharmacological studies. It functions as a respiratory stimulant analog and is used primarily as a stable-isotope internal standard for quantitative mass spectrometry and method development. The material is supplied as a high-purity solid suitable for laboratory handling.
  • Deuterium-labeled internal standard for quantitation.
  • High chemical purity and isotopic enrichment.
  • Solid, white to off-white, easy to handle and weigh.
  • Suitable for mass spectrometry and pharmacokinetic studies.
  • Available in small laboratory pack sizes.
Catalog No. 50-002-98918 Supplier Medchemexpress LLC Supplier No. HYB0551S10MG
Doxapram-d8 is a deuterium-labeled form of doxapram provided as a research standard for analytical and pharmacological studies. It functions as a respiratory stimulant analog and is used primarily as a stable-isotope internal standard for quantitative mass spectrometry and method development. The material is supplied as a high-purity solid suitable for laboratory handling.
  • Deuterium-labeled internal standard for quantitation.
  • High chemical purity and isotopic enrichment.
  • Solid, white to off-white, easy to handle and weigh.
  • Suitable for mass spectrometry and pharmacokinetic studies.
  • Available in small laboratory pack sizes.