Learn More
Selleck Chemical LLC Doxapram HCl-50mg
SDP
Supplier:  Selleck Chemical LLC S4037
Encompass_Preferred
Doxapram HCl inhibits TASK-1, TASK-3, TASK-1/TASK-3 heterodimeric channel function with EC50 of 410 nM, 37 μM, 9 μM, respectively.
Catalog No. 50-238-9693