Learn More
Medchemexpress LLC Doxapram (hydrochloride hydrate) | 7081-53-0 | 99.3% | 1 ML
SDP
Catalog No. 5000409005
Encompass_Preferred
Doxapram hydrochloride hydrate is a respiratory stimulant that increases breathing rate and depth by acting on the brain's respiratory centers and peripheral chemoreceptors. It can be used for the study of respiratory depression such as post-anesthesia respiratory depression, chronic obstructive pulmonary disease, and apnea of prematurity.
  • Inhibits TASK-1, TASK-3, and TASK-1/TASK-3 heterodimeric channel function
  • Inhibits Ca²⁺-activated and Ca²⁺-independent potassium current in type I cells of the carotid body
  • Significantly prolongs the effective refractory period of the atrium
  • Has an anti-arrhythmic effect
Catalog No. 50-004-09005 Supplier Medchemexpress LLC Supplier No. HYB0551A10MM/1ML
Doxapram hydrochloride hydrate is a respiratory stimulant that increases breathing rate and depth by acting on the brain's respiratory centers and peripheral chemoreceptors. It can be used for the study of respiratory depression such as post-anesthesia respiratory depression, chronic obstructive pulmonary disease, and apnea of prematurity.
  • Inhibits TASK-1, TASK-3, and TASK-1/TASK-3 heterodimeric channel function
  • Inhibits Ca²⁺-activated and Ca²⁺-independent potassium current in type I cells of the carotid body
  • Significantly prolongs the effective refractory period of the atrium
  • Has an anti-arrhythmic effect