Learn More
Medchemexpress LLC Doxapram hydrochloride hydrate | 7081-53-0 | MFCD09954698 | 99.3% | 432.98 g/mol | C24H33ClN2O3 | 10 MG
SDP
Catalog No. 5000298917
Encompass_Preferred
Doxapram hydrochloride hydrate is a small-molecule respiratory stimulant and TASK-1 potassium channel inhibitor supplied as a solid hydrate for research use. It is provided with high purity and is available in small solid quantities and pre-made DMSO solutions for assay work.
  • High purity (99.3%).
  • Molecular weight 432.98 g/mol.
  • Supplied as a solid hydrate; white to off-white appearance.
  • Available in small pack sizes suitable for assay development.
  • Offered in solution formats (10 mM in DMSO) for immediate use.
Catalog No. 50-002-98917 Supplier Medchemexpress LLC Supplier No. HYB0551A10MG
Doxapram hydrochloride hydrate is a small-molecule respiratory stimulant and TASK-1 potassium channel inhibitor supplied as a solid hydrate for research use. It is provided with high purity and is available in small solid quantities and pre-made DMSO solutions for assay work.
  • High purity (99.3%).
  • Molecular weight 432.98 g/mol.
  • Supplied as a solid hydrate; white to off-white appearance.
  • Available in small pack sizes suitable for assay development.
  • Offered in solution formats (10 mM in DMSO) for immediate use.