Learn More
Cayman Chemical Doxapram (hydrochloride hydrate), Purity: ≥95%, CAS: 7081-53-0, 25 mg
SDP
Catalog No. 502995228
Encompass_Preferred
A central respiratory stimulant; increases respiration and phrenic nerve activity in anesthetized cats at a dose of 1.0 mg/kg; acts through carotid chemoreceptors, lacking activity in anesthetized cats following carotid denervation
Catalog No. 50-299-5228 Supplier Cayman Chemical Supplier No. 2379425 MG
A central respiratory stimulant; increases respiration and phrenic nerve activity in anesthetized cats at a dose of 1.0 mg/kg; acts through carotid chemoreceptors, lacking activity in anesthetized cats following carotid denervation