Learn More
TARGETMOL CHEMICALS INC Doxazosin 50MG
SDP
Catalog No. 502876257
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Doxazosin (UK 33274) is a long-acting alpha1-adrenoceptor inhibitor widely used to treat benign prostatic hyperplasia and lower urinary tract symptoms. Purity 99.85%
Catalog No. 50-287-6257 Supplier TARGETMOL CHEMICALS INC Supplier No. T2231650MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Doxazosin (UK 33274) is a long-acting alpha1-adrenoceptor inhibitor widely used to treat benign prostatic hyperplasia and lower urinary tract symptoms. Purity 99.85%