Learn More
Apexbio Technology LLC Doxazosin 74191-85-8 500mg
SDP
Catalog No. 5000088095
Encompass_Preferred
Doxazosin (CAS 74191-85-8) is a synthetic quinazoline derivative that functions as a selective antagonist of postsynaptic 1-adrenergic receptors By inhibiting these receptors doxazosin reduces vascular smooth muscle contraction leading to decreased peripheral resistance This pharmacological profile is valuable for research investigating 1-adrenergic signaling pathways vascular tone regulation and cardiovascular physiology Doxazosin is commonly utilized in studies related to hypertension benign prostatic hyperplasia and receptor pharmacology
Catalog No. 50-000-88095 Supplier Apexbio Technology LLC Supplier No. B1209500
Doxazosin (CAS 74191-85-8) is a synthetic quinazoline derivative that functions as a selective antagonist of postsynaptic 1-adrenergic receptors By inhibiting these receptors doxazosin reduces vascular smooth muscle contraction leading to decreased peripheral resistance This pharmacological profile is valuable for research investigating 1-adrenergic signaling pathways vascular tone regulation and cardiovascular physiology Doxazosin is commonly utilized in studies related to hypertension benign prostatic hyperplasia and receptor pharmacology