Learn More
Medchemexpress LLC Doxazosin | 74191-85-8 | 99.9% | C23H25N5O5 | 25 MG
SDP
Catalog No. 5000359078
Encompass_Preferred
Doxazosin is a quinazoline-derivative that selectively antagonizes postsynaptic α1-adrenergic receptors. It is a long-lasting inhibitor of α1-adrenoceptors used to treat benign prostatic hyperplasia and lower urinary tract symptoms.
  • Selectively antagonizes postsynaptic α1-adrenergic receptors.
  • Long-lasting inhibitor of α1-adrenoceptors.
  • Used to treat benign prostatic hyperplasia.
  • Used to treat lower urinary tract symptoms.
  • May inhibit cholesterol synthesis and reduce LDL cholesterol.
Catalog No. 50-003-59078 Supplier Medchemexpress LLC Supplier No. 25MGHYB0098
Doxazosin is a quinazoline-derivative that selectively antagonizes postsynaptic α1-adrenergic receptors. It is a long-lasting inhibitor of α1-adrenoceptors used to treat benign prostatic hyperplasia and lower urinary tract symptoms.
  • Selectively antagonizes postsynaptic α1-adrenergic receptors.
  • Long-lasting inhibitor of α1-adrenoceptors.
  • Used to treat benign prostatic hyperplasia.
  • Used to treat lower urinary tract symptoms.
  • May inhibit cholesterol synthesis and reduce LDL cholesterol.