Learn More
Medchemexpress LLC Doxazosin | 74191-85-8 | MFCD00800482 | 99.9% | 451.48 | C23H25N5O5 | 10 MG
SDP
Catalog No. 5000298942
Encompass_Preferred
Doxazosin is a quinazoline-derived selective antagonist of postsynaptic α1-adrenergic receptors used in pharmacological research. Supplied as a white to off-white solid and available as a DMSO solution, it is provided for research use with defined purity and molecular identity.
  • Selective α1-adrenergic receptor antagonist for pharmacology studies.
  • Quinazoline derivative with defined chemical identity (CAS 74191-85-8).
  • Available as a solid and as a DMSO solution for assay flexibility.
  • High purity suitable for research applications.
  • Molecular formula C23H25N5O5 and molecular weight 451.48 g/mol.
Catalog No. 50-002-98942 Supplier Medchemexpress LLC Supplier No. HYB009810MG
Doxazosin is a quinazoline-derived selective antagonist of postsynaptic α1-adrenergic receptors used in pharmacological research. Supplied as a white to off-white solid and available as a DMSO solution, it is provided for research use with defined purity and molecular identity.
  • Selective α1-adrenergic receptor antagonist for pharmacology studies.
  • Quinazoline derivative with defined chemical identity (CAS 74191-85-8).
  • Available as a solid and as a DMSO solution for assay flexibility.
  • High purity suitable for research applications.
  • Molecular formula C23H25N5O5 and molecular weight 451.48 g/mol.